siRNA Details
| Gene Symbol: KCNQ1DN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024627.1 | 1 | 452 | |||
| GenBank mRNA | AB039920.1 | - | 1 | 452 | ||
| AJ404617.2 | - | 1 | 422 | |||
| BC098159.1 | - | 1 | 314 | |||
| BC098256.1 | - | 1 | 314 | |||
| BC098319.1 | - | 1 | 318 | |||
| BC098344.1 | - | 1 | 314 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000441418.2 | - | 1 | 452 | Chr. 11 - 2870033 - 2872105 [+] on Build GRCh38 | |

KCNQ1DN
Genomic Map