siRNA Details
| Gene Symbol: TFAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001288.1 | 1 | 587 | |||
| GenBank mRNA | CV358951.1 | - | 1 | 211 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TFAMP1| Gene Symbol: TFAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001288.1 | 1 | 587 | |||
| GenBank mRNA | CV358951.1 | - | 1 | 211 | ||