siRNA Details
| Gene Symbol: GGT8P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003503.1 | 2 | 2194 | |||
| GenBank mRNA | AK055838.1 | - | 2 | 2194 | ||
| Gene Symbol: GGT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005261890.4 | XP_005261947.1 | 2 | 261 | ||
| XM_017029183.1 | XP_016884672.1 | 2 | 237 | |||
| XM_017029185.1 | XP_016884674.1 | 8 | 2490 | |||
| Gene Symbol: LOC102724197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006724928.3 | XP_006724991.1 | 1 | 450 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV661840.1 | - | 1 | 256 | Chr. 22 - 24621494 - 24622041 [+] on Build GRCh38 | |

GGT8P
Genomic Map