siRNA Details
| Gene Symbol: LOC100129034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027406.1 | 1 | 3715 | |||
| GenBank mRNA | BC042968.1 | - | 1 | 243 | ||
| CR627469.1 | - | 1 | 1491 | |||
| Gene Symbol: PSMB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056654.1 | - | 1 | 1338 | ||

LOC100129034
Genomic Map