siRNA Details
| Gene Symbol: LOC100129034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027406.1 | 1 | 892 | |||
| GenBank mRNA | AK090539.1 | - | 1 | 96 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA234012.1 | - | 1 | 356 | Chr. 9 - 124354008 - 124354589 [+] on Build GRCh38 | |

LOC100129034
Genomic Map