siRNA Details
| Gene Symbol: LOC100129034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027406.1 | 1 | 2738 | |||
| GenBank mRNA | CR627469.1 | - | 1 | 514 | ||
| Gene Symbol: PSMB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056654.1 | - | 1 | 361 | ||

LOC100129034
Genomic Map