siRNA Details
| Gene Symbol: ASMTL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026710.1 | 2 | 153 | |||
| NR_026711.1 | 2 | 153 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASMTL-AS1| Gene Symbol: ASMTL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026710.1 | 2 | 153 | |||
| NR_026711.1 | 2 | 153 | ||||