siRNA Details
Gene Symbol: LOC153910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027311.1 | 3 | 1847 | |||
NR_027312.1 | 3 | 1850 | ||||
GenBank mRNA | AK055933.1 | - | 3 | 1850 | ||
BC033340.1 | - | 1 | 1169 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC153910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027311.1 | 3 | 1847 | |||
NR_027312.1 | 3 | 1850 | ||||
GenBank mRNA | AK055933.1 | - | 3 | 1850 | ||
BC033340.1 | - | 1 | 1169 |