siRNA Details
| Gene Symbol: POMGNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243766.1 | NP_001230695.1 | 1 | 549 | ||
| GenBank mRNA | AK056186.1 | - | 1 | 549 | ||
| CR749700.1 | - | 1 | 535 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

POMGNT1| Gene Symbol: POMGNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243766.1 | NP_001230695.1 | 1 | 549 | ||
| GenBank mRNA | AK056186.1 | - | 1 | 549 | ||
| CR749700.1 | - | 1 | 535 | |||