siRNA Details
| Gene Symbol: TBC1D3P1-DHX40P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002924.3 | 5 | 460 | |||
| GenBank mRNA | BX280208.1 | - | 5 | 383 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI828567.1 | - | 5 | 383 | Chr. 17 - 59995951 - 60018975 [-] on Build GRCh38 | |

TBC1D3P1-DHX40P1
Genomic Map