siRNA Details
| Gene Symbol: CEP170P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003135.2 | 1, 2 | 43 | |||
| GenBank mRNA | BC014590.2 | - | 1, 2 | 43 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEP170P1| Gene Symbol: CEP170P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003135.2 | 1, 2 | 43 | |||
| GenBank mRNA | BC014590.2 | - | 1, 2 | 43 | ||