siRNA Details
| Gene Symbol: LINC00200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015376.2 | 3 | 341 | |||
| GenBank mRNA | AK097673.1 | - | 3 | 291 | ||
| BC133038.1 | - | 3 | 341 | |||
| BC133040.1 | - | 3 | 293 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB045405.1 | - | 3 | 291 | Chr. 10 - 1159821 - 1162846 [+] on Build GRCh38 | |

LINC00200
Genomic Map