siRNA Details
| Gene Symbol: LINC00200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015376.2 | 3 | 2189 | |||
| GenBank mRNA | AK097673.1 | - | 3 | 2183 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00200| Gene Symbol: LINC00200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015376.2 | 3 | 2189 | |||
| GenBank mRNA | AK097673.1 | - | 3 | 2183 | ||