siRNA Details
Gene Symbol: LINC00999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024497.2 | 8 | 1638 | |||
GenBank mRNA | AK128746.1 | - | 1 | 195 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024497.2 | 8 | 1638 | |||
GenBank mRNA | AK128746.1 | - | 1 | 195 |