siRNA Details
| Gene Symbol: LINC00999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024497.2 | 8 | 2078 | |||
| GenBank mRNA | AK128746.1 | - | 1 | 635 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00999| Gene Symbol: LINC00999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024497.2 | 8 | 2078 | |||
| GenBank mRNA | AK128746.1 | - | 1 | 635 | ||