siRNA Details
| Gene Symbol: LOC285074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026846.1 | 10, 11 | 1965 | |||
| GenBank mRNA | BC069048.1 | - | 10, 11 | 1747 | ||
| Gene Symbol: LOC730268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017005464.1 | XP_016860953.1 | 13, 14 | 1771 | ||

LOC285074
Genomic Map