siRNA Details
| Gene Symbol: LINC00264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026793.1 | 1 | 47 | |||
| GenBank mRNA | BC038537.1 | - | 1 | 47 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00264| Gene Symbol: LINC00264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026793.1 | 1 | 47 | |||
| GenBank mRNA | BC038537.1 | - | 1 | 47 | ||