siRNA Details
| Gene Symbol: LINC00924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027132.1 | 7 | 2319 | |||
| NR_027133.1 | 6 | 2232 | ||||
| GenBank mRNA | AK057337.1 | - | 7 | 2321 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00924| Gene Symbol: LINC00924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027132.1 | 7 | 2319 | |||
| NR_027133.1 | 6 | 2232 | ||||
| GenBank mRNA | AK057337.1 | - | 7 | 2321 | ||