siRNA Details
| Gene Symbol: LINC00924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027132.1 | 4 | 587 | |||
| NR_027133.1 | 4 | 587 | ||||
| GenBank mRNA | AK057337.1 | - | 4 | 587 | ||
| BC131607.1 | - | 4 | 586 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00924| Gene Symbol: LINC00924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027132.1 | 4 | 587 | |||
| NR_027133.1 | 4 | 587 | ||||
| GenBank mRNA | AK057337.1 | - | 4 | 587 | ||
| BC131607.1 | - | 4 | 586 | |||