siRNA Details
| Gene Symbol: EFHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243252.1 | NP_001230181.1 | 1, 2 | 110 | ||
| GenBank mRNA | AK310565.1 | - | 1, 2 | 110 | ||
| DB461847.1 | - | 1, 2 | 107 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EFHD1| Gene Symbol: EFHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243252.1 | NP_001230181.1 | 1, 2 | 110 | ||
| GenBank mRNA | AK310565.1 | - | 1, 2 | 110 | ||
| DB461847.1 | - | 1, 2 | 107 | |||