siRNA Details
| Gene Symbol: FER1L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_119376.1 | 34 | 4251 | |||
| GenBank mRNA | AK023521.1 | - | 352 | |||
| AL833764.1 | - | 670 | ||||
| AY461812.1 | - | 34 | 4418 | |||
| BX648024.1 | - | 8 | 1074 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FER1L4| Gene Symbol: FER1L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_119376.1 | 34 | 4251 | |||
| GenBank mRNA | AK023521.1 | - | 352 | |||
| AL833764.1 | - | 670 | ||||
| AY461812.1 | - | 34 | 4418 | |||
| BX648024.1 | - | 8 | 1074 | |||