siRNA Details
| Gene Symbol: FER1L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_119376.1 | 9 | 799 | |||
| GenBank mRNA | AY461812.1 | - | 9 | 733 | ||
| BC029951.1 | - | 9 | 725 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FER1L4| Gene Symbol: FER1L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_119376.1 | 9 | 799 | |||
| GenBank mRNA | AY461812.1 | - | 9 | 733 | ||
| BC029951.1 | - | 9 | 725 | |||