siRNA Details
| Gene Symbol: LRRC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001309474.1 | NP_001296403.1 | 1 | 180 | ||
| XM_011540210.2 | XP_011538512.1 | 1 | 178 | |||
| GenBank mRNA | DC352688.1 | - | 1 | 181 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC27| Gene Symbol: LRRC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001309474.1 | NP_001296403.1 | 1 | 180 | ||
| XM_011540210.2 | XP_011538512.1 | 1 | 178 | |||
| GenBank mRNA | DC352688.1 | - | 1 | 181 | ||