siRNA Details
| Gene Symbol: LRRC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001309474.1 | NP_001296403.1 | 11 | |||
| GenBank mRNA | DC352688.1 | - | 12 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX436744.2 | - | 1 | 64 | Chr. 10 - 132337054 - 132337904 [+] on Build GRCh38 | |

LRRC27
Genomic Map