siRNA Details
| Gene Symbol: LRRC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001309474.1 | NP_001296403.1 | 10 | 1611 | ||
| XM_006717984.2 | XP_006718047.1 | 11 | 1701 | |||
| GenBank mRNA | AK301422.1 | - | 10 | 1378 | ||
| BC058159.1 | - | 5 | 588 | |||

LRRC27
Genomic Map