siRNA Details
| Gene Symbol: PPP1R2P9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002191.2 | 1 | 265 | |||
| GenBank mRNA | AB044137.1 | - | 1 | 265 | ||
| BC056673.1 | - | 1 | 195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1R2P9| Gene Symbol: PPP1R2P9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002191.2 | 1 | 265 | |||
| GenBank mRNA | AB044137.1 | - | 1 | 265 | ||
| BC056673.1 | - | 1 | 195 | |||