siRNA Details
| Gene Symbol: ZNF667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321356.1 | NP_001308285.1 | 3 | 585 | ||
| XM_011527208.1 | XP_011525510.1 | 2 | 2229 | |||
| GenBank mRNA | AK096513.1 | - | 3 | 514 | ||
| AK126957.1 | - | 1 | 1224 | |||
| AK127426.1 | - | 2 | 2229 | |||

ZNF667
Genomic Map