siRNA Details
| Gene Symbol: LOC399815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027282.1 | 2, 3 | 534 | |||
| GenBank mRNA | AK300776.1 | - | 2, 3 | 438 | ||
| BC053499.1 | - | 4, 5 | 779 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA905937.1 | - | 2, 3 | 447 | Chr. 10 - 122879730 - 122884782 [+] on Build GRCh38 | |

LOC399815
Genomic Map