siRNA Details
| Gene Symbol: LOC399815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027282.1 | 5 | 907 | |||
| GenBank mRNA | AK300776.1 | - | 5 | 811 | ||
| BC053499.1 | - | 7 | 1223 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC399815| Gene Symbol: LOC399815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027282.1 | 5 | 907 | |||
| GenBank mRNA | AK300776.1 | - | 5 | 811 | ||
| BC053499.1 | - | 7 | 1223 | |||