siRNA Details
| Gene Symbol: OTX2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029385.1 | 4 | 1182 | |||
| GenBank mRNA | AB593040.1 | - | 3 | 1172 | ||
| BC041486.2 | - | 4 | 1181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OTX2-AS1| Gene Symbol: OTX2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029385.1 | 4 | 1182 | |||
| GenBank mRNA | AB593040.1 | - | 3 | 1172 | ||
| BC041486.2 | - | 4 | 1181 | |||