siRNA Details
| Gene Symbol: SNAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030717.1 | 1 | 81 | |||
| GenBank mRNA | BP234322.1 | - | 1 | 80 | ||
| BX647416.1 | - | 1 | 48 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNAPC2| Gene Symbol: SNAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030717.1 | 1 | 81 | |||
| GenBank mRNA | BP234322.1 | - | 1 | 80 | ||
| BX647416.1 | - | 1 | 48 | |||