siRNA Details
Gene Symbol: CYP1B1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027252.1 | 3 | 1338 | |||
GenBank mRNA | BC031410.1 | - | 3 | 1338 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CYP1B1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027252.1 | 3 | 1338 | |||
GenBank mRNA | BC031410.1 | - | 3 | 1338 |