siRNA Details
| Gene Symbol: LINC01312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027030.1 | 3 | 2268 | |||
| GenBank mRNA | BC032958.2 | - | 3 | 2268 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01312| Gene Symbol: LINC01312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027030.1 | 3 | 2268 | |||
| GenBank mRNA | BC032958.2 | - | 3 | 2268 | ||