siRNA Details
| Gene Symbol: MIR17HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027349.1 | 4 | 714 | |||
| NR_027350.1 | 2 | 4805 | ||||
| GenBank mRNA | AK024240.1 | - | 1 | 2528 | ||
| Gene Symbol: MIR18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AB176708.1 | - | LIT3364 oncomir-1 |
2 | 4805 | |

MIR17HG
Genomic Map