siRNA Details
| Gene Symbol: WFDC21P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030732.1 | 1 | 235 | |||
| GenBank mRNA | AF086204.1 | - | 1 | 61 | ||
| KJ020271.1 | - | 1 | 83 | |||
| Gene Symbol: LOC105371849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BU630466.1 | - | 1 | 476 | ||

WFDC21P
Genomic Map