siRNA Details
| Gene Symbol: WFDC21P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030732.1 | 1, 2 | 264 | |||
| GenBank mRNA | AF086204.1 | - | 1, 2 | 90 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WFDC21P| Gene Symbol: WFDC21P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030732.1 | 1, 2 | 264 | |||
| GenBank mRNA | AF086204.1 | - | 1, 2 | 90 | ||