siRNA Details
| Gene Symbol: PPP2R2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033191.2 | 767 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP2R2D| Gene Symbol: PPP2R2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033191.2 | 767 | ||||