siRNA Details
| Gene Symbol: ASH1L-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027023.1 | 1 | 1719 | |||
| GenBank mRNA | AK126559.1 | - | 1 | 1670 | ||
| BC017347.2 | - | 2 | 664 | |||
| BC065229.1 | - | 1 | 440 | |||
| BC104640.1 | - | 1 | 458 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ940588.1 | - | 1 | 705 | Chr. 1 - 155562997 - 155563797 [+] on Build GRCh38 | |

ASH1L-AS1
Genomic Map