siRNA Details
| Gene Symbol: SPRR2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003062.1 | 2 | 255 | |||
| GenBank mRNA | BC120937.1 | - | 2 | 213 | ||
| M21539.1 | - | 2 | 255 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPRR2C| Gene Symbol: SPRR2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003062.1 | 2 | 255 | |||
| GenBank mRNA | BC120937.1 | - | 2 | 213 | ||
| M21539.1 | - | 2 | 255 | |||