siRNA Details
| Gene Symbol: RPL13AP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004844.1 | 1 | 739 | |||
| GenBank mRNA | BC067891.1 | - | 1 | 739 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL13AP3| Gene Symbol: RPL13AP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004844.1 | 1 | 739 | |||
| GenBank mRNA | BC067891.1 | - | 1 | 739 | ||