siRNA Details
Gene Symbol: RPL13AP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004844.1 | 1 | 980 | |||
GenBank mRNA | BC067891.1 | - | 1 | 980 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RPL13AP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004844.1 | 1 | 980 | |||
GenBank mRNA | BC067891.1 | - | 1 | 980 |