siRNA Details
| Gene Symbol: IQSEC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243197.1 | NP_001230126.1 | 3 | 861 | ||
| GenBank mRNA | BC038213.1 | - | 2 | 745 | ||
| BC108674.1 | - | 3 | 845 | |||
| BX119489.1 | - | 1 | 158 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IQSEC2| Gene Symbol: IQSEC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243197.1 | NP_001230126.1 | 3 | 861 | ||
| GenBank mRNA | BC038213.1 | - | 2 | 745 | ||
| BC108674.1 | - | 3 | 845 | |||
| BX119489.1 | - | 1 | 158 | |||