siRNA Details
| Gene Symbol: FAR2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026758.2 | 6 | 2332 | |||
| GenBank mRNA | AK125921.1 | - | 6 | 2334 | ||
| AL043002.1 | - | 3 | 357 | |||
| BC132694.1 | - | 1 | 30 | |||
| BC132696.1 | - | 1 | 30 | |||
| BC144438.1 | - | 1 | 30 | 9052961 | ||

FAR2P1
Genomic Map