siRNA Details
| Gene Symbol: FAR2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026758.2 | 12 | 3868 | |||
| Gene Symbol: FAR2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046258.1 | 6 | 2196 | |||
| NR_046259.1 | 5 | 2098 | ||||
| NR_046260.1 | 4 | 1973 | ||||
| GenBank mRNA | AK054990.1 | - | 4 | 1413 | ||

FAR2P1
Genomic Map