siRNA Details
| Gene Symbol: FAR2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046258.1 | 6 | 1895 | |||
| NR_046259.1 | 5 | 1797 | ||||
| NR_046260.1 | 4 | 1672 | ||||
| GenBank mRNA | AK054990.1 | - | 4 | 1112 | ||
| BC131487.1 | - | 2 | 3720 | |||
| Gene Symbol: FAR2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026758.2 | 12 | 3571 | |||
| GenBank mRNA | AK125921.1 | - | 12 | 3572 | ||

FAR2P2
Genomic Map