siRNA Details
| Gene Symbol: MISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291291.1 | NP_001278220.1 | 2 | 920 | ||
| NR_111941.1 | 2 | 985 | ||||
| GenBank mRNA | BC011002.2 | - | 2 | 920 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MISP3| Gene Symbol: MISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291291.1 | NP_001278220.1 | 2 | 920 | ||
| NR_111941.1 | 2 | 985 | ||||
| GenBank mRNA | BC011002.2 | - | 2 | 920 | ||