siRNA Details
Gene Symbol: MISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291291.1 | NP_001278220.1 | 3 | 997 | ||
NR_111941.1 | 3 | 1062 | ||||
GenBank mRNA | BC011002.2 | - | 3 | 997 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291291.1 | NP_001278220.1 | 3 | 997 | ||
NR_111941.1 | 3 | 1062 | ||||
GenBank mRNA | BC011002.2 | - | 3 | 997 |