siRNA Details
| Gene Symbol: MISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291291.1 | NP_001278220.1 | 3 | 1247 | ||
| NR_111941.1 | 3 | 1312 | ||||
| GenBank mRNA | BC011002.2 | - | 3 | 1247 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI088010.1 | - | 3 | 603 | Chr. 19 - 14073652 - 14075050 [+] on Build GRCh38 | |

MISP3
Genomic Map