siRNA Details
| Gene Symbol: LINC01124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027433.1 | 1 | 354 | |||
| GenBank mRNA | AK123485.1 | - | 1 | 354 | ||
| BC132926.1 | - | 1 | 311 | |||
| BC132928.1 | - | 1 | 311 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01124| Gene Symbol: LINC01124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027433.1 | 1 | 354 | |||
| GenBank mRNA | AK123485.1 | - | 1 | 354 | ||
| BC132926.1 | - | 1 | 311 | |||
| BC132928.1 | - | 1 | 311 | |||