siRNA Details
| Gene Symbol: GABPB1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024490.1 | 1 | 2114 | |||
| GenBank mRNA | BX537481.1 | - | 2 | 267 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GABPB1-AS1| Gene Symbol: GABPB1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024490.1 | 1 | 2114 | |||
| GenBank mRNA | BX537481.1 | - | 2 | 267 | ||